Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrothiophene-4-carbonitrile, 97%

Chemical Identifiers

Chemical Name or Material 2-Nitrothiophene-4-carbonitrile
CAS 42137-23-5
Molecular Formula C5H2N2O2S
Formula Weight 154.15
MDL Number MFCD00052580
UN Number UN3439
Beilstein 123937
Synonym 4-Cyano-2-nitrothiophene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1142803

  SDS
alfa aesar™
L1142803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL11428MD

  SDS
alfa aesar™
L11428MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.15
Chemical Name or Material 2-Nitrothiophene-4-carbonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Molecular Formula C5H2N2O2S
MDL Number MFCD00052580
CAS 42137-23-5
UN Number UN3439
Synonym 4-Cyano-2-nitrothiophene
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Melting Point 121° to 123°C
Quantity 1g
Assay 97%
Beilstein 123937

Applications
2-Nitrothiophene-4-carbonitrile is used as an intermediate in chemical research.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.