missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrothiophene-4-carbonitrile, 97%

Manufacturer:  Alfa Aesar™ L11428MD

 View more versions of this product

Catalog No. AAL11428MD



Description

Description

Applications
2-Nitrothiophene-4-carbonitrile is used as an intermediate in chemical research.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2-Nitrothiophene-4-carbonitrile
250mg
C5H2N2O2S
UN3439
2-nitrothiophene-4-carbonitrile, acmc-1aogv, maybridge1_003117, 2-nitro-4-cyanothiophene, 4-cyano-2-nitrothiophene, 5-nitro-3-thiophenecarbonitrile, nacmzlgghmpmkq-uhfffaoysa, 5-nitro-3-thiophenecarbonitrile #, 5-nitro-thiophene-3-carbonitrile, 5-nitro-3-thiophenecarbonitrile; 2-nitro-4-cyanothiophene; 2-cyano-4-nitrothiophene
C1=C(SC=C1C#N)[N+](=O)[O-]
154.143
154.15
121°C to 123°C
42137-23-5
MFCD00052580
123937
NACMZLGGHMPMKQ-UHFFFAOYSA-N
5-nitrothiophene-3-carbonitrile
142579
97%

Chemical Identifiers

42137-23-5
154.143
NACMZLGGHMPMKQ-UHFFFAOYSA-N
142579
C1=C(SC=C1C#N)[N+](=O)[O-]
C5H2N2O2S
MFCD00052580
2-nitrothiophene-4-carbonitrile, acmc-1aogv, maybridge1_003117, 2-nitro-4-cyanothiophene, 4-cyano-2-nitrothiophene, 5-nitro-3-thiophenecarbonitrile, nacmzlgghmpmkq-uhfffaoysa, 5-nitro-3-thiophenecarbonitrile #, 5-nitro-thiophene-3-carbonitrile, 5-nitro-3-thiophenecarbonitrile; 2-nitro-4-cyanothiophene; 2-cyano-4-nitrothiophene
5-nitrothiophene-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents