missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitropropane 95.0+%, TCI America™

Supplier:  TCI America N024925G

 View more versions of this product

Catalog No. N024925G



Description

Description

Specifications

Specifications

2-Nitropropane
-91°C
120°C
MFCD00007397
2608
FGLBSLMDCBOPQK-UHFFFAOYSA-N
2-nitropropane
398
89.09
Liquid
79-46-9
Yellow
C3H7NO2
25 g
propane, 2-nitro, isonitropropane, dimethylnitromethane, nitroisopropane, sec-nitropropane, nipar s-20, beta-nitropropane, 2-np, nipar s-20 solvent, nipar s-30 solvent
CC(C)[N+](=O)[O-]
89.094
CHEBI:16037
≥95.0% (GC)

Chemical Identifiers

79-46-9
89.094
FGLBSLMDCBOPQK-UHFFFAOYSA-N
398
2-nitropropane
C3H7NO2
MFCD00007397
propane, 2-nitro, isonitropropane, dimethylnitromethane, nitroisopropane, sec-nitropropane, nipar s-20, beta-nitropropane, 2-np, nipar s-20 solvent, nipar s-30 solvent
CHEBI:16037
CC(C)[N+](=O)[O-]
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0194

RTECSNumber : TZ5250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions