Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrophloroglucinol, 98%

$330.95 - $1,446.92

Chemical Identifiers

Chemical Name or Material 2-Nitrophloroglucinol
CAS 16600-92-3
Molecular Formula C6H5NO5
Formula Weight 171.11
MDL Number MFCD00016996
Beilstein 1963331
Synonym 1-Nitro-2,4,6-trihydroxybenzene; 2,4,6-Trihydroxynitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1369406

  SDS
alfa aesar™
A1369406
5g Each for $330.95

AAA1369414

  SDS
alfa aesar™
A1369414
25g Each for $1,446.92
Description & Specifications

Specifications

Formula Weight 171.11
Chemical Name or Material 2-Nitrophloroglucinol
Assay 98%
Molecular Formula C6H5NO5
MDL Number MFCD00016996
Melting Point 187° to 191°C
CAS 16600-92-3
Quantity 5g
Beilstein 1963331
Synonym 1-Nitro-2,4,6-trihydroxybenzene; 2,4,6-Trihydroxynitrobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.