missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrophenylarsonic Acid, TCI America™

$35.14 - $204.72

Chemical Identifiers

CAS 5410-29-7
Molecular Formula C6H6AsNO5
Molecular Weight (g/mol) 247.038
MDL Number MFCD00047664
InChI Key UYEDGVZDGVIURN-UHFFFAOYSA-N
Synonym 2-nitrobenzenearsonic acid, arsonic acid, 2-nitrophenyl, o-nitrophenylarsenic acid, o-nitrobenzenearsonic acid, o-nitrophenyl arsonic acid, benzenearsonic acid, o-nitro, unii-hnr3su2y5p, 2-nitrophenyl arsonic acid, arsonic acid, as-2-nitrophenyl, hnr3su2y5p
PubChem CID 79424
IUPAC Name (2-nitrophenyl)arsonic acid
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])[As](=O)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N01331G View Documents TCI America
N01331G
1g Each for $35.14
 
N013325G View Documents TCI America
N013325G
25g Each for $204.72
 
Specifications

Chemical Identifiers

5410-29-7
247.038
UYEDGVZDGVIURN-UHFFFAOYSA-N
79424
C1=CC=C(C(=C1)[N+](=O)[O-])[As](=O)(O)O
C6H6AsNO5
MFCD00047664
2-nitrobenzenearsonic acid, arsonic acid, 2-nitrophenyl, o-nitrophenylarsenic acid, o-nitrobenzenearsonic acid, o-nitrophenyl arsonic acid, benzenearsonic acid, o-nitro, unii-hnr3su2y5p, 2-nitrophenyl arsonic acid, arsonic acid, as-2-nitrophenyl, hnr3su2y5p
(2-nitrophenyl)arsonic acid

Specifications

Undesignated
5410-29-7
MFCD00047664
2-nitrobenzenearsonic acid, arsonic acid, 2-nitrophenyl, o-nitrophenylarsenic acid, o-nitrobenzenearsonic acid, o-nitrophenyl arsonic acid, benzenearsonic acid, o-nitro, unii-hnr3su2y5p, 2-nitrophenyl arsonic acid, arsonic acid, as-2-nitrophenyl, hnr3su2y5p
C1=CC=C(C(=C1)[N+](=O)[O-])[As](=O)(O)O
247.038
247.04
≥98.0% (T)
1g
C6H6AsNO5
3465
UYEDGVZDGVIURN-UHFFFAOYSA-N
(2-nitrophenyl)arsonic acid
79424
Crystal-Powder at 20°C
2-Nitrophenylarsonic Acid
Safety and Handling

Safety and Handling

RTECSNumber : CY5950000

TSCA : Yes