Please login to your online account to display your discounted pricing

2-nitrophenylarsonic acid, +90%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC376930050

 View more versions of this product

Catalog No. AC376930050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥90.0%
Physical Form Powder
Formula Weight 247.04
Quantity 5g
Color Beige-yellow
CAS Min % 90.0
CAS Max % 100.0
Assay Percent Range 90+%
Linear Formula NO2C6H4AsO3H2
MDL Number MFCD00047664
CAS 5410-29-7
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Infrared Spectrum Authentic
Chemical Name or Material 2-Nitrophenylarsonic acid
Molecular Formula C6H6AsNO5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Nitrophenylarsonic acid
CAS 5410-29-7
CAS Min % 90.0
CAS Max % 100.0
Assay ≥90.0%
Assay Percent Range 90+%
Molecular Formula C6H6AsNO5
Linear Formula NO2C6H4AsO3H2
Formula Weight 247.04
MDL Number MFCD00047664
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

SDS & Safety Documents