Please login to your online account to display your discounted pricing

2-nitrophenylarsonic acid, +90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Nitrophenylarsonic acid
CAS 5410-29-7
CAS Min % 90.0
CAS Max % 100.0
Assay ≥90.0%
Assay Percent Range 90+%
Molecular Formula C6H6AsNO5
Linear Formula NO2C6H4AsO3H2
Formula Weight 247.04
MDL Number MFCD00047664
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376930010

  SDS
thermo fisher scientific
AC376930010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376930050

  SDS
thermo fisher scientific
AC376930050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥90.0%
Formula Weight 247.04
Physical Form Powder
Quantity 1g
Color Beige-yellow
CAS Max % 100.0
CAS Min % 90.0
Linear Formula NO2C6H4AsO3H2
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 2-Nitrophenylarsonic acid
MDL Number MFCD00047664
Assay Percent Range 90+%
Infrared Spectrum Authentic
Molecular Formula C6H6AsNO5
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 5410-29-7
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.