Learn More
2-Nitrophenylacetonitrile, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 610-66-2 |
---|---|
Molecular Formula | C8H6N2O2 |
Molecular Weight (g/mol) | 162.15 |
MDL Number | MFCD00007183 |
InChI Key | YPRFCQAWSNWRLM-UHFFFAOYSA-N |
Synonym | 2-2-nitrophenyl acetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl cyanide, 2-nitrobenzyl nitrile, 2-nitrophenyl acetonitrile, 2-nitrophenylacetonitrile, acetonitrile, o-nitrophenyl, benzeneacetonitrile, 2-nitro, o-nitrobenzacetonitrile, o-nitrophenyl acetonitrile |
PubChem CID | 11888 |
IUPAC Name | 2-(2-nitrophenyl)acetonitrile |
SMILES | [O-][N+](=O)C1=CC=CC=C1CC#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC172420100
|
Thermo Fisher Scientific
172420100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC172420500
|
Thermo Fisher Scientific
AC172420500 |
50g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
610-66-2 | |
162.15 | |
YPRFCQAWSNWRLM-UHFFFAOYSA-N | |
11888 | |
[O-][N+](=O)C1=CC=CC=C1CC#N |
C8H6N2O2 | |
MFCD00007183 | |
2-2-nitrophenyl acetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl cyanide, 2-nitrobenzyl nitrile, 2-nitrophenyl acetonitrile, 2-nitrophenylacetonitrile, acetonitrile, o-nitrophenyl, benzeneacetonitrile, 2-nitro, o-nitrobenzacetonitrile, o-nitrophenyl acetonitrile | |
2-(2-nitrophenyl)acetonitrile |
Specifications
610-66-2 | |
100.0 | |
81.0°C to 85.0°C | |
Authentic | |
O2NC6H4CH2CN | |
09, 455 | |
2-2-nitrophenyl acetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl cyanide, 2-nitrobenzyl nitrile, 2-nitrophenyl acetonitrile, 2-nitrophenylacetonitrile, acetonitrile, o-nitrophenyl, benzeneacetonitrile, 2-nitro, o-nitrobenzacetonitrile, o-nitrophenyl acetonitrile | |
YPRFCQAWSNWRLM-UHFFFAOYSA-N | |
2-(2-nitrophenyl)acetonitrile | |
11888 | |
Powder | |
2-Nitrophenylacetonitrile |
97.5 | |
Brown to Brown-Gray | |
97.5% min. (HPLC) | |
C8H6N2O2 | |
MFCD00007183 | |
Glass bottle | |
10g | |
[O-][N+](=O)C1=CC=CC=C1CC#N | |
162.15 | |
162.15 | |
98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
EINECSNumber : 210-231-
RUO – Research Use Only