Learn More
2-Nitrophenylacetonitrile, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 172420100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Nitrophenylacetonitrile | |
97.5 | |
Authentic | |
Glass bottle | |
O2NC6H4CH2CN | |
10g | |
2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl | |
[O-][N+](=O)C1=CC=CC=C1CC#N | |
162.15 | |
162.15 |
610-66-2 | |
100.0 | |
97.5% min. (HPLC) | |
C8H6N2O2 | |
MFCD00007183 | |
09, 455 | |
YPRFCQAWSNWRLM-UHFFFAOYSA-N | |
2-(2-nitrophenyl)acetonitrile | |
11888 | |
98% |
Chemical Identifiers
610-66-2 | |
162.15 | |
YPRFCQAWSNWRLM-UHFFFAOYSA-N | |
11888 | |
[O-][N+](=O)C1=CC=CC=C1CC#N |
C8H6N2O2 | |
MFCD00007183 | |
2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl | |
2-(2-nitrophenyl)acetonitrile |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
EINECSNumber : 210-231-
RUO â Research Use Only