Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrophenyl phenyl ether, 99%

$57.57 - $898.03

Chemical Identifiers

Chemical Name or Material 2-Nitrophenyl phenyl ether
CAS 2216-12-8
Molecular Formula C12H9NO3
Formula Weight 215.21
MDL Number MFCD00035744
Beilstein 2051914
Synonym 2-Nitrodiphenyl ether; 2-Phenoxynitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1808809

  SDS
alfa aesar™
A1808809
10g Each for $57.57

AAA1808818

  SDS
alfa aesar™
A1808818
50g Each for $215.47

AAA1808830

  SDS
alfa aesar™
A1808830
250g Each for $898.03
Description & Specifications

Specifications

Formula Weight 215.21
Molecular Formula C12H9NO3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Nitrophenyl phenyl ether
MDL Number MFCD00035744
Synonym 2-Nitrodiphenyl ether; 2-Phenoxynitrobenzene
Health Hazard 1 Warning
CAS 2216-12-8
TSCA TSCA
Assay 99%
Flash Point 181°C (358°F)
Boiling Point 184°C (8mmHg)
Beilstein 2051914
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
RTECS Number KO2650000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.