Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrophenyl isothiocyanate, 97%

$102.68 - $102.68

Chemical Identifiers

Chemical Name or Material 2-Nitrophenyl isothiocyanate
CAS 2719-30-4
Molecular Formula C7H4N2O2S
Formula Weight 180.19
MDL Number MFCD00007092
UN Number UN2923
Beilstein 2719657
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0966906

  SDS
alfa aesar™
L0966906*
5g Each for $102.68
Description & Specifications

Specifications

Formula Weight 180.19
Chemical Name or Material 2-Nitrophenyl isothiocyanate
Assay 97%
Molecular Formula C7H4N2O2S
MDL Number MFCD00007092
CAS 2719-30-4
UN Number UN2923
Beilstein 2719657
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Flash Point >110°C (230°F)
Odor Irritating
Boiling Point 135° to 137°C (17mmHg)
Melting Point 73° to 76°C
Quantity 5g

Applications
2-Nitrophenyl isothiocyanate can react with pyridin-2-ylamine to get 2-nitrophenyl(2-pyridyl)thiourea. It is also used as pharmaceutical intermediates.

Solubility
Hydrolyzes with water.

Notes
Store away from moisture and oxidizing agents. Moisture Sensitive. Protect from heat. Incompatible with oxidizing agents, amines, alcohols and water.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.