Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrophenyl beta-D-glucopyranoside, 98+%

$88.05 - $349.42

Chemical Identifiers

Chemical Name or Material 2-Nitrophenyl beta-D-glucopyranoside
CAS 2816-24-2
Molecular Formula C12H15NO8
Formula Weight 301.25
MDL Number MFCD00065048
Beilstein 92206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1493503

 
alfa aesar™
A1493503
1g Each for $88.05

AAA1493506

 
alfa aesar™
A1493506
5g Each for $349.42
Description & Specifications

Specifications

Formula Weight 301.25
Chemical Name or Material 2-Nitrophenyl beta-D-glucopyranoside
Assay ≥98%
Molecular Formula C12H15NO8
MDL Number MFCD00065048
CAS 2816-24-2
Beilstein 92206
Recommended Storage 2° to 8°C
Optical Rotation −105° (c=2 in Water)
Melting Point 141° to 142°C
Quantity 1g

Applications
2-Nitrophenyl beta-D-glucopyranoside is used as a substrate for ß -glucosidase.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

2° to 8°C