Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrophenoxyacetic acid, 98+%

$34.49 - $489.49

Chemical Identifiers

Chemical Name or Material 2-Nitrophenoxyacetic acid
CAS 1878-87-1
Molecular Formula C8H7NO5
Formula Weight 197.15
MDL Number MFCD00016993
Beilstein 2114909
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1622606

 
alfa aesar™
A1622606
5g Each for $34.49

AAA1622614

 
alfa aesar™
A1622614
25g Each for $143.78

AAA1622622

 
alfa aesar™
A1622622
100g Each for $489.49
Description & Specifications

Specifications

Formula Weight 197.15
Chemical Name or Material 2-Nitrophenoxyacetic acid
Assay ≥98%
Molecular Formula C8H7NO5
MDL Number MFCD00016993
CAS 1878-87-1
Beilstein 2114909
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 156° to 159°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.