Learn More
2-Nitrophenol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 128740050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
IndicatorSpecifications
2-Nitrophenol | |
88-75-5 | |
100.0 | |
108°C | |
98.5% min. (GC) | |
C6H5NO3 | |
MFCD00011688 | |
1663 | |
05,477 | |
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech | |
IQUPABOKLQSFBK-UHFFFAOYSA-N | |
2-nitrophenol | |
6947 | |
CHEBI:16260 | |
99% |
99% | |
98.5 | |
214.0°C to 216.0°C | |
Authentic | |
Glass bottle | |
O2NC6H4OH | |
5g | |
06, 213 | |
15, 6707 | |
Solubility in water: 2g/L (25°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene,carbon disulfide,alkali hydroxides,and toluene | |
OC1=CC=CC=C1[N+]([O-])=O | |
139.11 | |
1.35 mPa.s (80°C) | |
139.11 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
GHS Signal Word: Warning
EINECSNumber : 201-857-5
RTECSNumber : SM2100000
TSCA : TSCA
RUO â Research Use Only