missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrophenol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1233330

 View more versions of this product

Catalog No. AAA1233330



Description

Description

2-Nitrophenol is a building block used as an acid-base indicator and corrosion inhibitor for aluminum-copper alloy. It is also used to make dyes, paint coloring, rubber chemicals, and substances that kill molds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrophenol is a building block used as an acid-base indicator and corrosion inhibitor for aluminum-copper alloy. It is also used to make dyes, paint coloring, rubber chemicals, and substances that kill molds.

Solubility
Soluble in water (2.1 g/L at 20°C), ethanol, chloroform, alcohol, and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications

Specifications

2-Nitrophenol
44°C to 48°C
214°C to 216°C
Phenol-like
MFCD00011688
UN1663
14,6620
Soluble in water (2.1g/L at 20°C),ethanol,chloroform,alcohol,and acetone.
OC1=CC=CC=C1[N+]([O-])=O
139.11
CHEBI:16260
98%
88-75-5
1.26
108°C (226°F)
C6H5NO3
250 g
775403
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
IQUPABOKLQSFBK-UHFFFAOYSA-N
2-nitrophenol
6947
139.11

Chemical Identifiers

88-75-5
139.11
IQUPABOKLQSFBK-UHFFFAOYSA-N
6947
2-nitrophenol
C6H5NO3
MFCD00011688
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
CHEBI:16260
OC1=CC=CC=C1[N+]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
H373-H302
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS

EINECSNumber : 201-857-5

RTECSNumber : SM2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only