missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrofluorenone, TCI America™

$82.50 - $404.88

Chemical Identifiers

CAS 3096-52-4
Molecular Formula C13H7NO3
Molecular Weight (g/mol) 225.203
MDL Number MFCD00001152
InChI Key AJEAHBZZHSLIQP-UHFFFAOYSA-N
Synonym 2-nitro-9h-fluoren-9-one, 2-nitro-9-fluorenone, 2-nitrofluorenone, 9h-fluoren-9-one, 2-nitro, ccris 2540, 9-fluorenone, 2-nitro, 4-07-00-01636 beilstein handbook reference, 2-nitro-9-fluorenone, 2-nitro-9-fluorenone, 2-nitro-9h-fluoren-9-one #
PubChem CID 18356
IUPAC Name 2-nitrofluoren-9-one
SMILES C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N02021G View Documents TCI America
N02021G
1g Each for $82.50
 
N020210G View Documents TCI America
N020210G
10g Each for $404.88
 
Specifications

Chemical Identifiers

3096-52-4
225.203
AJEAHBZZHSLIQP-UHFFFAOYSA-N
18356
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)[N+](=O)[O-]
C13H7NO3
MFCD00001152
2-nitro-9h-fluoren-9-one, 2-nitro-9-fluorenone, 2-nitrofluorenone, 9h-fluoren-9-one, 2-nitro, ccris 2540, 9-fluorenone, 2-nitro, 4-07-00-01636 beilstein handbook reference, 2-nitro-9-fluorenone, 2-nitro-9-fluorenone, 2-nitro-9h-fluoren-9-one #
2-nitrofluoren-9-one

Specifications

Yellow
1g
C13H7NO3
2-nitro-9h-fluoren-9-one, 2-nitro-9-fluorenone, 2-nitrofluorenone, 9h-fluoren-9-one, 2-nitro, ccris 2540, 9-fluorenone, 2-nitro, 4-07-00-01636 beilstein handbook reference, 2-nitro-9-fluorenone, 2-nitro-9-fluorenone, 2-nitro-9h-fluoren-9-one #
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)[N+](=O)[O-]
225.203
225.20
≥98.0% (HPLC,N)
222°C
3096-52-4
MFCD00001152
AJEAHBZZHSLIQP-UHFFFAOYSA-N
2-nitrofluoren-9-one
18356
Crystal-Powder at 20°C
2-Nitrofluorenone
Safety and Handling

Safety and Handling

RTECSNumber : LL9091000

TSCA : No