Learn More
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
$57.59 - $206.15
Chemical Identifiers
CAS | 607-57-8 |
---|---|
Molecular Formula | C13H9NO2 |
Molecular Weight (g/mol) | 211.22 |
MDL Number | MFCD00001117 |
InChI Key | XFOHWECQTFIEIX-UHFFFAOYSA-N |
Synonym | 2-nitrofluorene, nitrofluorene, 9h-fluorene, 2-nitro, fluorene, 2-nitro, unii-191ll4u4gz, ccris 1189, dsstox_cid_971, dsstox_rid_75898, dsstox_gsid_20971 |
PubChem CID | 11831 |
ChEBI | CHEBI:1224 |
IUPAC Name | 2-nitro-9H-fluorene |
SMILES | [O-][N+](=O)C1=CC=C2C(CC3=CC=CC=C23)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5574406
|
Thermo Scientific Chemicals
H5574406 |
5 g |
Each for $57.59
|
|
|||||
AAH5574414
|
Thermo Scientific Chemicals
H5574414 |
25 g |
Each for $206.15
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
607-57-8 | |
211.22 | |
XFOHWECQTFIEIX-UHFFFAOYSA-N | |
11831 | |
2-nitro-9H-fluorene |
C13H9NO2 | |
MFCD00001117 | |
2-nitrofluorene, nitrofluorene, 9h-fluorene, 2-nitro, fluorene, 2-nitro, unii-191ll4u4gz, ccris 1189, dsstox_cid_971, dsstox_rid_75898, dsstox_gsid_20971 | |
CHEBI:1224 | |
[O-][N+](=O)C1=CC=C2C(CC3=CC=CC=C23)=C1 |
Specifications
607-57-8 | |
White to Brown | |
MFCD00001117 | |
UN3077 | |
XFOHWECQTFIEIX-UHFFFAOYSA-N | |
2-nitro-9H-fluorene | |
11831 | |
211.22 | |
Powder |
156°C to 158°C | |
C13H9NO2 | |
5 g | |
2-nitrofluorene, nitrofluorene, 9h-fluorene, 2-nitro, fluorene, 2-nitro, unii-191ll4u4gz, ccris 1189, dsstox_cid_971, dsstox_rid_75898, dsstox_gsid_20971 | |
[O-][N+](=O)C1=CC=C2C(CC3=CC=CC=C23)=C1 | |
211.22 | |
CHEBI:1224 | |
98% | |
2-Nitrofluorene |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P281-P308+P313-P501c
H341-H351
DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 210-138-5
RTECSNumber : LL8225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only