missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrofluorene, 98%

Manufacturer:  Alfa Aesar™ H5574414

 View more versions of this product

Catalog No. AAH5574414



Specifications

Specifications

2-Nitrofluorene
156°C to 158°C
607-57-8
MFCD00001117
2-nitrofluorene
C1C2=CC=CC=C2C3=C1C=C(C=C3)[N+](=O)[O-]
211.22
CHEBI:1224
Powder
White to Brown
25g
C13H9NO2
UN3077
XFOHWECQTFIEIX-UHFFFAOYSA-N
2-nitro-9H-fluorene
11831
211.22
98%

Chemical Identifiers

607-57-8
211.22
XFOHWECQTFIEIX-UHFFFAOYSA-N
11831
2-nitro-9H-fluorene
C13H9NO2
MFCD00001117
2-nitrofluorene
CHEBI:1224
C1C2=CC=CC=C2C3=C1C=C(C=C3)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : LL8225000

TSCA : TSCA

Documents