missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrocinnamaldehyde, predominantly trans, 98%

$64.00 - $229.77

Chemical Identifiers

CAS 1466-88-2
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00007188
InChI Key VMSMELHEXDVEDE-HWKANZROSA-N
Synonym 2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
PubChem CID 5367122
IUPAC Name (E)-3-(2-nitrophenyl)prop-2-enal
SMILES C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1030814 View Documents Alfa Aesar™
A1030814
25g Each for $127.23
 
AAA1030818 View Documents Alfa Aesar™
A1030818
50g Each for $229.77
 
AAA1030809 View Documents Alfa Aesar™
A1030809
10g Each for $64.00
 
Description

Description

Applications
It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

1466-88-2
177.159
VMSMELHEXDVEDE-HWKANZROSA-N
5367122
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
C9H7NO3
MFCD00007188
2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
(E)-3-(2-nitrophenyl)prop-2-enal

Specifications

Characteristic
25g
C9H7NO3
908376
VMSMELHEXDVEDE-HWKANZROSA-N
(E)-3-(2-nitrophenyl)prop-2-enal
5367122
98%
125°C to 129°C
1466-88-2
MFCD00007188
2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
177.159
177.16
2-Nitrocinnamaldehyde, predominantly trans
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning