Learn More
2-Nitrocinnamaldehyde, predominantly trans, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1030818
Description
It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
2-Nitrocinnamaldehyde | |
1466-88-2 | |
Characteristic | |
MFCD00007188 | |
908376 | |
2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775 | |
VMSMELHEXDVEDE-HWKANZROSA-N | |
(E)-3-(2-nitrophenyl)prop-2-enal | |
5367122 | |
98% |
predominantly trans | |
125°C to 129°C | |
C9H7NO3 | |
50 g | |
Air Sensitive | |
Soluble in methanol. Insoluble in water. | |
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-] | |
177.159 | |
177.16 |
Chemical Identifiers
1466-88-2 | |
177.159 | |
VMSMELHEXDVEDE-HWKANZROSA-N | |
5367122 | |
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-] |
C9H7NO3 | |
MFCD00007188 | |
2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775 | |
(E)-3-(2-nitrophenyl)prop-2-enal |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 215-988-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only