missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1890003

 View more versions of this product

Catalog No. AAA1890003



Description

Description

2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies. Also reactant involved in synthesis of tetrahydroisoquinolines via Heck-aza-Michael reactions, aminomethylphenylamine scaffolds for factor Xa inhibition, squaryliamides for mannosyltransferase inhibition and fluorescent two-photon absorption benzothiadiazole dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzylamine hydrochloride may be used in chemical synthesis studies. Also reactant involved in synthesis of tetrahydroisoquinolines via Heck-aza-Michael reactions, aminomethylphenylamine scaffolds for factor Xa inhibition, squaryliamides for mannosyltransferase inhibition and fluorescent two-photon absorption benzothiadiazole dyes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

2-Nitrobenzylamine hydrochloride
250°C (decomposition)
MFCD00136280
Hygroscopic
BASJTVIZZDEQBJ-UHFFFAOYSA-N
(2-nitrophenyl)methanamine;hydrochloride
12235442
98%
24835-08-3
C7H9ClN2O2
1 g
2-nitrobenzylamine hydrochloride, 2-nitrophenyl methanamine hydrochloride, 1-2-nitrophenyl methanamine hydrochloride, 2-nitrobenzylamine hcl, 2-nitrobenzyl amine hcl, acmc-1co7n, o-nitrobenzylamine hydrochloride, 2-nitrobenzyl amine hydrochloride, 2-nitrobenzenemethanamine hydrochloride, 2-nitrophenyl methanaminehydrochloride
C1=CC=C(C(=C1)CN)[N+](=O)[O-].Cl
188.611
188.62

Chemical Identifiers

24835-08-3
188.611
BASJTVIZZDEQBJ-UHFFFAOYSA-N
12235442
C1=CC=C(C(=C1)CN)[N+](=O)[O-].Cl
C7H9ClN2O2
MFCD00136280
2-nitrobenzylamine hydrochloride, 2-nitrophenyl methanamine hydrochloride, 1-2-nitrophenyl methanamine hydrochloride, 2-nitrobenzylamine hcl, 2-nitrobenzyl amine hcl, acmc-1co7n, o-nitrobenzylamine hydrochloride, 2-nitrobenzyl amine hydrochloride, 2-nitrobenzenemethanamine hydrochloride, 2-nitrophenyl methanaminehydrochloride
(2-nitrophenyl)methanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only