Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Nitrobenzyl)triphenylphosphonium bromide monohydrate, 98+%

$131.46 - $518.74

Chemical Identifiers

Chemical Name or Material (2-Nitrobenzyl)triphenylphosphonium bromide monohydrate
CAS 23308-83-0
Molecular Formula C25H21BrNO2P·H2O
Formula Weight 496.35 (478.33 Anhydrous)
MDL Number MFCD04971966
Beilstein 4086172
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0565004

  SDS
alfa aesar™
L0565004
2g Each for $131.46

AAL0565009

  SDS
alfa aesar™
L0565009
10g Each for $518.74
Description & Specifications

Specifications

Formula Weight 496.35 (478.33 Anhydrous)
Assay ≥98%
Molecular Formula C25H21BrNO2P·H2O
MDL Number MFCD04971966
Chemical Name or Material (2-Nitrobenzyl)triphenylphosphonium bromide monohydrate
Beilstein 4086172
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 2g
CAS 23308-83-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈236° to 240°C

Applications
(2-Nitrobenzyl)triphenylphosphonium bromide monohydrate is used as intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.