missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrobenzyl Chloride, TCI America™

$96.25 - $271.95

Chemical Identifiers

CAS 612-23-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007185
InChI Key BXCBUWKTXLWPSB-UHFFFAOYSA-N
Synonym 2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
PubChem CID 11921
IUPAC Name 1-(chloromethyl)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)CCl)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N01825G View Documents TCI America
N01825G
5g Each for $96.25
 
N018225G View Documents TCI America
N018225G
25g Each for $271.95
 
Specifications

Chemical Identifiers

612-23-7
171.58
BXCBUWKTXLWPSB-UHFFFAOYSA-N
11921
C1=CC=C(C(=C1)CCl)[N+](=O)[O-]
C7H6ClNO2
MFCD00007185
2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
1-(chloromethyl)-2-nitrobenzene

Specifications

133°C
49°C
612-23-7
MFCD00007185
2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
C1=CC=C(C(=C1)CCl)[N+](=O)[O-]
171.58
171.58
≥98.0% (GC,T)
Yellow
5g
C7H6ClNO2
3261
BXCBUWKTXLWPSB-UHFFFAOYSA-N
1-(chloromethyl)-2-nitrobenzene
11921
Crystal-Powder at 20°C
2-Nitrobenzyl Chloride
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0449

RTECSNumber : XS9092000

TSCA : No