missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrobenzyl chloride, 98+%

Manufacturer:  Alfa Aesar™ L1359914

 View more versions of this product

Catalog No. AAL1359914



Description

Description

Applications
2-Nitrobenzyl chloride has been used to introduce the photo labile 2-nitrobenzyl protecting group for the phenolic OH group of tyrosine. It is used as a pharmaceutical intermediate.

Solubility
Soluble in acetone, alcohol benzene, diethyl ether, and ethanol (98%). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

2-Nitrobenzyl chloride
127°C to 133°C (10mmHg)
25g
C7H6ClNO2
UN3261
2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
C1=CC=C(C(=C1)CCl)[N+](=O)[O-]
171.58
171.58
>110°C (230°F)
47°C to 51°C
612-23-7
MFCD00007185
388396
BXCBUWKTXLWPSB-UHFFFAOYSA-N
1-(chloromethyl)-2-nitrobenzene
11921
≥98%

Chemical Identifiers

612-23-7
171.58
BXCBUWKTXLWPSB-UHFFFAOYSA-N
11921
C1=CC=C(C(=C1)CCl)[N+](=O)[O-]
C7H6ClNO2
MFCD00007185
2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
1-(chloromethyl)-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H341
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : XS9092000

Documents