Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrobenzyl chloride, 98+%

Chemical Identifiers

Chemical Name or Material 2-Nitrobenzyl chloride
CAS 612-23-7
Molecular Formula C7H6ClNO2
Formula Weight 171.58
MDL Number MFCD00007185
UN Number UN3261
Beilstein 388396
Synonym alpha-Chloro-2-nitrotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1359906

  SDS
alfa aesar™
L1359906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1359914

  SDS
alfa aesar™
L1359914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.58
Chemical Name or Material 2-Nitrobenzyl chloride
CAS 612-23-7
Molecular Formula C7H6ClNO2
UN Number UN3261
MDL Number MFCD00007185
Synonym alpha-Chloro-2-nitrotoluene
Health Hazard 2 GHS H Statement
H314-H318-H341
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 127° to 133°C (10mmHg)
Beilstein 388396
Quantity 5g
Melting Point 47° to 51°C
Assay ≥98%
RTECS Number XS9092000

Applications
2-Nitrobenzyl chloride has been used to introduce the photo labile 2-nitrobenzyl protecting group for the phenolic OH group of tyrosine. It is used as a pharmaceutical intermediate.

Solubility
Soluble in acetone, alcohol benzene, diethyl ether, and ethanol (98%). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H341
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.