missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrobenzoyl chloride, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC232311000

 View more versions of this product

Catalog No. AC232311000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Nitrobenzoylchloride
Authentic
1.5666 to 1.5686
148°C to 149°C (9.0mmHg)
17°C to 20°C
610-14-0
100.0
C7H4ClNO3
MFCD00007131
Solubility in water: decomposes
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-]
185.57
185.57
95%
1.4000g/mL
Glass bottle
1.4
Brown to Yellow
100g
94.0
95%
O2NC6H4COCl
2-nitro-benzoyl chloride, 2-nitrobenzoylchloride, benzoyl chloride, 2-nitro, benzoyl chloride, nitro, benzoyl chloride, o-nitro, ccris 3135, dsstox_cid_5741, nitrobenzoyl chloride, o-nitrobenzoyl chloride, unii-86w2g0ja5q
BWWHTIHDQBHTHP-UHFFFAOYSA-N
2-nitrobenzoyl chloride
11875
Liquid After Melting

Chemical Identifiers

610-14-0
185.57
BWWHTIHDQBHTHP-UHFFFAOYSA-N
11875
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-]
C7H4ClNO3
MFCD00007131
2-nitro-benzoyl chloride, 2-nitrobenzoylchloride, benzoyl chloride, 2-nitro, benzoyl chloride, nitro, benzoyl chloride, o-nitro, ccris 3135, dsstox_cid_5741, nitrobenzoyl chloride, o-nitrobenzoyl chloride, unii-86w2g0ja5q
2-nitrobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-206-4

SDS Documents