missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrobenzoyl chloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 232310250

 View more versions of this product

Catalog No. AC232310250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Nitrobenzoyl chloride
94.0
17°C to 20°C
1.4000g/mL
Authentic
Glass bottle
1.5666 to 1.5686
MFCD00007131
1.4
Solubility in water: decomposes
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-]
185.57
185.57
Liquid After Melting
610-14-0
100.0
Brown to Yellow
148°C to 149°C (9.0 mmHg)
95%
C7H4ClNO3
O2NC6H4COCl
25 g
o-nitrobenzoyl chloride, benzoyl chloride, 2-nitro, 2-nitrobenzoylchloride, benzoyl chloride, o-nitro, unii-86w2g0ja5q, nitrobenzoyl chloride, ccris 3135, 2-nitro-benzoyl chloride, benzoyl chloride, nitro, dsstox_cid_5741
BWWHTIHDQBHTHP-UHFFFAOYSA-N
2-nitrobenzoyl chloride
11875
95%

Chemical Identifiers

610-14-0
185.57
BWWHTIHDQBHTHP-UHFFFAOYSA-N
11875
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-]
C7H4ClNO3
MFCD00007131
o-nitrobenzoyl chloride, benzoyl chloride, 2-nitro, 2-nitrobenzoylchloride, benzoyl chloride, o-nitro, unii-86w2g0ja5q, nitrobenzoyl chloride, ccris 3135, 2-nitro-benzoyl chloride, benzoyl chloride, nitro, dsstox_cid_5741
2-nitrobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-206-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only