Learn More
2-Nitrobenzoyl chloride, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 232310050
Specifications
2-Nitrobenzoyl chloride | |
Authentic | |
1.5666 to 1.5686 | |
148°C to 149°C (9.0mmHg) | |
17°C to 20°C | |
610-14-0 | |
100.0 | |
C7H4ClNO3 | |
MFCD00007131 | |
Solubility in water: decomposes | |
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-] | |
185.57 | |
185.57 | |
95% |
1.4000g/mL | |
Glass bottle | |
1.4 | |
Brown to Yellow | |
5g | |
94.0 | |
95% | |
O2NC6H4COCl | |
2-nitro-benzoyl chloride, 2-nitrobenzoylchloride, benzoyl chloride, 2-nitro, benzoyl chloride, nitro, benzoyl chloride, o-nitro, ccris 3135, dsstox_cid_5741, nitrobenzoyl chloride, o-nitrobenzoyl chloride, unii-86w2g0ja5q | |
BWWHTIHDQBHTHP-UHFFFAOYSA-N | |
2-nitrobenzoyl chloride | |
11875 | |
Liquid After Melting |
Chemical Identifiers
610-14-0 | |
185.57 | |
BWWHTIHDQBHTHP-UHFFFAOYSA-N | |
11875 | |
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-] |
C7H4ClNO3 | |
MFCD00007131 | |
2-nitro-benzoyl chloride, 2-nitrobenzoylchloride, benzoyl chloride, 2-nitro, benzoyl chloride, nitro, benzoyl chloride, o-nitro, ccris 3135, dsstox_cid_5741, nitrobenzoyl chloride, o-nitrobenzoyl chloride, unii-86w2g0ja5q | |
2-nitrobenzoyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 210-206-4