missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrobenzotrifluoride, 98%, Acros Organics™

Chemical Identifiers

CAS 384-22-5
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
InChI Key NDZJSUCUYPZXPR-UHFFFAOYSA-N
Synonym 1-nitro-2-trifluoromethyl benzene
PubChem CID 9795
IUPAC Name 1-nitro-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC166520010 SDS View Documents Thermo Fisher Scientific
AC166520010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC166520050 View Documents Thermo Fisher Scientific
AC166520050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

384-22-5
191.11
1-nitro-2-trifluoromethyl benzene
1-nitro-2-(trifluoromethyl)benzene
C7H4F3NO2
NDZJSUCUYPZXPR-UHFFFAOYSA-N
9795
C1=CC=C(C(=C1)C(F)(F)F)[N+](=O)[O-]

Specifications

95°C
Glass Bottle
31.0°C to 32.0°C
384-22-5
C7H4F3NO2
05,II,251
NDZJSUCUYPZXPR-UHFFFAOYSA-N
1-nitro-2-(trifluoromethyl)benzene
9795
98%
Authentic
216.0°C to 217.0°C
1g
97.5% min. (GC)
O2NC6H4CF3
1-nitro-2-trifluoromethyl benzene
C1=CC=C(C(=C1)C(F)(F)F)[N+](=O)[O-]
191.11
191.11
2-Nitrobenzotrifluoride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated cl

GHS Signal Word: Danger

EINECSNumber : 206-855-8