missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrobenzonitrile, TCI America™

$51.95 - $137.50

Chemical Identifiers

CAS 612-24-8
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.121
MDL Number MFCD00007044
InChI Key SWBDKCMOLSUXRH-UHFFFAOYSA-N
Synonym o-nitrobenzonitrile, o-cyanonitrobenzene, benzonitrile, o-nitro, benzonitrile, 2-nitro, 2-cyanonitrobenzene, nitrobenzonitrile, unii-drc6u29fci, ccris 2326, 1-nitro-2-cyanobenzene, 2-nitrobenzenecarbonitrile
PubChem CID 11922
IUPAC Name 2-nitrobenzonitrile
SMILES C1=CC=C(C(=C1)C#N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N017125G View Documents TCI America
N017125G
25g Each for $137.50
 
N01715G View Documents TCI America
N01715G
5g Each for $51.95
 
Specifications

Chemical Identifiers

612-24-8
148.121
SWBDKCMOLSUXRH-UHFFFAOYSA-N
11922
C1=CC=C(C(=C1)C#N)[N+](=O)[O-]
C7H4N2O2
MFCD00007044
o-nitrobenzonitrile, o-cyanonitrobenzene, benzonitrile, o-nitro, benzonitrile, 2-nitro, 2-cyanonitrobenzene, nitrobenzonitrile, unii-drc6u29fci, ccris 2326, 1-nitro-2-cyanobenzene, 2-nitrobenzenecarbonitrile
2-nitrobenzonitrile

Specifications

165°C
110°C
612-24-8
MFCD00007044
o-nitrobenzonitrile, o-cyanonitrobenzene, benzonitrile, o-nitro, benzonitrile, 2-nitro, 2-cyanonitrobenzene, nitrobenzonitrile, unii-drc6u29fci, ccris 2326, 1-nitro-2-cyanobenzene, 2-nitrobenzenecarbonitrile
C1=CC=C(C(=C1)C#N)[N+](=O)[O-]
148.121
148.12
≥98.0% (GC)
Yellow
25g
C7H4N2O2
3439
SWBDKCMOLSUXRH-UHFFFAOYSA-N
2-nitrobenzonitrile
11922
Crystal-Powder at 20°C
2-Nitrobenzonitrile
Safety and Handling

Safety and Handling

RTECSNumber : DI4903000

TSCA : No