Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrobenzonitrile, 98+%

Chemical Identifiers

Chemical Name or Material 2-Nitrobenzonitrile
CAS 612-24-8
Molecular Formula C7H4N2O2
Formula Weight 148.12
MDL Number MFCD00007044
UN Number UN3439
Beilstein 638987
Synonym 2-Cyanonitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1094106

  SDS
alfa aesar™
A1094106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.12
Odor Almond-like
Health Hazard 3 GHS P Statement
P280h-P309-P310-P302+P352
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Melting Point 107° to 111°C
Molecular Formula C7H4N2O2
MDL Number MFCD00007044
Quantity 5g
Beilstein 638987
Assay ≥98%
RTECS Number DI4903000
CAS 612-24-8
Health Hazard 2 GHS H Statement
H300-H311
Fatal if swallowed.
Toxic in contact with skin.
Synonym 2-Cyanonitrobenzene
Chemical Name or Material 2-Nitrobenzonitrile
UN Number UN3439
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Boiling Point 165°C (16mmHg)

Applications
ET catalysis of Au25 in intramolecular cascade reaction of 2-nitrobenzonitrile, from which 2-amniobenzamide (a precursor of significant pharmaceuticals20,21) is produced with high yield. fluorodenitration of 2-nitrobenzonitrile with Rb18F in DMSO gave an 85% yield of the 18F-labelled benzonitrile after only 20 minutes at 150°C.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P309-P310-P302+P352
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H300-H311
Fatal if swallowed.
Toxic in contact with skin.