Learn More
2-Nitrobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 128431000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Nitrobenzenesulfonyl chloride | |
63.0°C to 67.0°C | |
Glass bottle | |
SO2ClC6H4NO2 | |
100 g | |
06,419 | |
WPHUUIODWRNJLO-UHFFFAOYSA-N | |
2-nitrobenzenesulfonyl chloride | |
15540 | |
97% |
1694-92-4 | |
96% min. (GC) | |
C6H4ClNO4S | |
MFCD00007430 | |
11, 67 | |
2-nitrobenzene-1-sulfonyl chloride, o-nitrobenzenesulfonyl chloride, o-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, 2-nitro, 2-nitrobenzenesulphonyl chloride, 2-nitrobenzenesulfonylchloride, 2-nitro-benzenesulfonyl chloride, benzenesulfonyl chloride, o-nitro, 2-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, nitro | |
C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)Cl | |
221.61 | |
221.61 |
Chemical Identifiers
1694-92-4 | |
221.61 | |
WPHUUIODWRNJLO-UHFFFAOYSA-N | |
15540 | |
C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)Cl |
C6H4ClNO4S | |
MFCD00007430 | |
2-nitrobenzene-1-sulfonyl chloride, o-nitrobenzenesulfonyl chloride, o-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, 2-nitro, 2-nitrobenzenesulphonyl chloride, 2-nitrobenzenesulfonylchloride, 2-nitro-benzenesulfonyl chloride, benzenesulfonyl chloride, o-nitro, 2-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, nitro | |
2-nitrobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 216-907-1
RUO – Research Use Only