missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrobenzenesulfonic Acid Hydrate, TCI America™

$71.81 - $215.42

Chemical Identifiers

CAS 80-82-0
Molecular Formula C6H5NO5S
Molecular Weight (g/mol) 203.168
MDL Number MFCD00053617
InChI Key HWTDMFJYBAURQR-UHFFFAOYSA-N
PubChem CID 6664
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N01395G View Documents TCI America
N01395G
5g Each for $71.81
 
N013925G View Documents TCI America
N013925G
25g Each for $215.42
 
Specifications

Chemical Identifiers

80-82-0
203.168
HWTDMFJYBAURQR-UHFFFAOYSA-N
C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)O
C6H5NO5S
MFCD00053617
6664

Specifications

Yellow
80-82-0
MFCD00053617
HWTDMFJYBAURQR-UHFFFAOYSA-N
203.168
203.17
≥98.0% (HPLC,T)
5g
C6H5NO5S
2305
C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)O
6664
Crystal-Powder at 20°C
2-Nitrobenzenesulfonic Acid Hydrate
Safety and Handling

Safety and Handling

TSCA : No