missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrobenzenesulfenyl chloride, 98%, Acros Organics™

Manufacturer:  Thermo Fisher Scientific 161330500

 View more versions of this product

Catalog No. AC161330500



Specifications

Specifications

2-Nitrobenzenesulfenyl chloride
Glass bottle
70.0°C to 75.0°C
99%
98.5
97.5% min. (HPLC)
O2NC6H4SCl
06, I, 157
2-nitrobenzenesulfenyl chloride
NTNKNFHIAFDCSJ-UHFFFAOYSA-N
(2-nitrophenyl) thiohypochlorite
24319
Crystalline Powder or Needles
Authentic
Golden to Yellow
50g
7669-54-7
100.0
C6H4ClNO2S
MFCD00007128
01,745; 04,359; 06,419; 08,360; 10,277; 15,217
Solubility in water: decomposes
C1=CC=C(C(=C1)[N+](=O)[O-])SCl
189.62
189.62
98%

Chemical Identifiers

7669-54-7
189.62
NTNKNFHIAFDCSJ-UHFFFAOYSA-N
24319
C1=CC=C(C(=C1)[N+](=O)[O-])SCl
C6H4ClNO2S
MFCD00007128
2-nitrobenzenesulfenyl chloride
(2-nitrophenyl) thiohypochlorite
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-644-2

TSCA : TSCA

SDS Documents