missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrobenzaldoxime, 98+%

Manufacturer:  Alfa Aesar™ A1456506

 View more versions of this product

Catalog No. AAA1456506



Specifications

Specifications

2-Nitrobenzaldoxime
5g
C7H6N2O3
UN2811
2-nitrobenzaldoxime, 2-nitrobenzaldehyde oxime, o-nitrobenzaldoxime, benzaldehyde, 2-nitro-, oxime, e-n-hydroxy-1-2-nitrophenyl methanimine, ne-n-2-nitrophenyl methylidene hydroxylamine, syn-o-nitrobenzaldoxime, e-n-2-nitrophenyl methylidene hydroxylamine, 2-nitrobenzaldehyde oxime #, 1e-2-nitrobenzaldehyde oxime
C1=CC=C(C(=C1)C=NO)[N+](=O)[O-]
166.136
166.14
98°C to 102°C
6635-41-2
MFCD00007187
2614466
IHMGDCCTWRRUDX-VMPITWQZSA-N
(NE)-N-[(2-nitrophenyl)methylidene]hydroxylamine
7332808
≥98%

Chemical Identifiers

6635-41-2
166.136
IHMGDCCTWRRUDX-VMPITWQZSA-N
7332808
C1=CC=C(C(=C1)C=NO)[N+](=O)[O-]
C7H6N2O3
MFCD00007187
2-nitrobenzaldoxime, 2-nitrobenzaldehyde oxime, o-nitrobenzaldoxime, benzaldehyde, 2-nitro-, oxime, e-n-hydroxy-1-2-nitrophenyl methanimine, ne-n-2-nitrophenyl methylidene hydroxylamine, syn-o-nitrobenzaldoxime, e-n-2-nitrophenyl methylidene hydroxylamine, 2-nitrobenzaldehyde oxime #, 1e-2-nitrobenzaldehyde oxime
(NE)-N-[(2-nitrophenyl)methylidene]hydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents