Learn More
2-Nitrobenzaldehyde, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 128405000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Nitrobenzaldehyde | |
153.0°C (23.0 mmHg) | |
Authentic | |
Plastic bottle | |
O2NC6H4CHO | |
07,243 | |
o-nitrobenzaldehyde, benzaldehyde, 2-nitro, nitrobenzaldehyde, benzaldehyde, o-nitro, 2-nitro-benzaldehyde, 2-formylnitrobenzene, ccris 2322, unii-48b18q9b8e, benzaldehyde, nitro, ortho-nitrobenzaldehyde | |
CMWKITSNTDAEDT-UHFFFAOYSA-N | |
2-nitrobenzaldehyde | |
11101 | |
151.12 |
552-89-6 | |
>110°C | |
99% min. (GC) | |
C7H5NO3 | |
500g | |
14,6587 | |
Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol,benzene and ether | |
C1=CC=C(C(=C1)C=O)[N+](=O)[O-] | |
151.12 | |
CHEBI:66927 | |
99+% |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for s
GHS Signal Word: Warning
EINECSNumber : 209-025-3
RUO â Research Use Only