Learn More
2-Nitroaniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A110550B
Description
2-Nitroaniline is the main precursor to phenylenediamines, which are converted to benzimidazoles, a family of heterocycles that are key components in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Nitroaniline is the main precursor to phenylenediamines, which are converted to benzimidazoles, a family of heterocycles that are key components in pharmaceuticals.
Solubility
Soluble in water (0.6 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2-Nitroaniline | |
70°C to 74°C | |
284°C to 286°C | |
C6H6N2O2 | |
1000 g | |
509275 | |
o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzene | |
DPJCXCZTLWNFOH-UHFFFAOYSA-N | |
2-nitroaniline | |
6946 | |
98% |
88-74-4 | |
1.255 | |
168°C (334°F) | |
MFCD00007687 | |
UN1661 | |
14,6583 | |
Soluble in water (0.6g/L at 20°C). | |
C1=CC=C(C(=C1)N)[N+](=O)[O-] | |
138.126 | |
138.13 |
Chemical Identifiers
88-74-4 | |
138.126 | |
DPJCXCZTLWNFOH-UHFFFAOYSA-N | |
6946 | |
C1=CC=C(C(=C1)N)[N+](=O)[O-] |
C6H6N2O2 | |
MFCD00007687 | |
o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzene | |
2-nitroaniline |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROANILINES
EINECSNumber : 201-855-4
RTECSNumber : BY6650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only