missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitro-p-xylylene Glycol, TCI America™

$145.14 - $436.95

Chemical Identifiers

CAS 23222-97-1
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.163
MDL Number MFCD00024286
InChI Key KWVHOBYJXDKIPL-UHFFFAOYSA-N
PubChem CID 90035
IUPAC Name [4-(hydroxymethyl)-3-nitrophenyl]methanol
SMILES C1=CC(=C(C=C1CO)[N+](=O)[O-])CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N05191G View Documents TCI America
N05191G
1g Each for $145.14
 
N05195G View Documents TCI America
N05195G
5g Each for $436.95
 
Specifications

Chemical Identifiers

23222-97-1
183.163
KWVHOBYJXDKIPL-UHFFFAOYSA-N
[4-(hydroxymethyl)-3-nitrophenyl]methanol
C8H9NO4
MFCD00024286
90035
C1=CC(=C(C=C1CO)[N+](=O)[O-])CO

Specifications

Undesignated
1g
C8H9NO4
KWVHOBYJXDKIPL-UHFFFAOYSA-N
[4-(hydroxymethyl)-3-nitrophenyl]methanol
90035
Crystal-Powder at 20°C
2-Nitro-p-xylylene Glycol
95°C
23222-97-1
MFCD00024286
C1=CC(=C(C=C1CO)[N+](=O)[O-])CO
183.163
183.16
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No