Please login to your online account to display your discounted pricing

2-Nitro-p-phenylenediamine 95-99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  128801000

 View more versions of this product

Catalog No. AC128801000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Formula Weight 153.14
Physical Form Powder
Quantity 100g
Color Brown to black
Beilstein 13, 120
RTECS Number ST3000000
TSCA TSCA
EINECS Number 226-164-5
Melting Point 134° to 140°C
CAS Max % 100.0
CAS Min % 95.0
Synonym 1, 4-Diamino-2-nitrobenzene
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95-99%
Name Note 95-99%
Molecular Formula C6H7N3O2
Linear Formula O2NC6H3(NH2)2
CAS 5307-14-2
Chemical Name or Material 2-Nitro-p-phenylenediamine
MDL Number MFCD00007903

Specifications

Chemical Name or Material 2-Nitro-p-phenylenediamine
Name Note 95-99%
CAS 5307-14-2
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 95-99%
Molecular Formula C6H7N3O2
Linear Formula O2NC6H3(NH2)2
Formula Weight 153.14
MDL Number MFCD00007903
Synonym 1, 4-Diamino-2-nitrobenzene
Accessories

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.

SDS & Safety Documents