Please login to your online account to display your discounted pricing

2-Nitro-p-phenylenediamine 95-99%, ACROS Organics™

$89.57 - $89.57

Chemical Identifiers

Chemical Name or Material 2-Nitro-p-phenylenediamine
Name Note 95-99%
CAS 5307-14-2
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 95-99%
Molecular Formula C6H7N3O2
Linear Formula O2NC6H3(NH2)2
Formula Weight 153.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC128801000

  SDS
thermo fisher scientific
128801000
Glass bottle 100g Each for $89.57

AC128805000

  SDS
thermo fisher scientific
128805000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Powder
Formula Weight 153.14
Quantity 100g
Color Brown to black
Beilstein 13, 120
EINECS Number 226-164-5
TSCA TSCA
Melting Point 134° to 140°C
RTECS Number ST3000000
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2-Nitro-p-phenylenediamine
Packaging Glass bottle
Name Note 95-99%
Assay Percent Range 95-99%
Molecular Formula C6H7N3O2
Linear Formula O2NC6H3(NH2)2
MDL Number MFCD00007903
CAS 5307-14-2
Synonym 1, 4-Diamino-2-nitrobenzene
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.