Please login to your online account to display your discounted pricing

2-Nitro-p-phenylenediamine 95-99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  128805000

 View more versions of this product

Catalog No. AC128805000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Powder
Formula Weight 153.14
Quantity 500g
Color Brown to black
EINECS Number 226-164-5
TSCA TSCA
Melting Point 134° to 140°C
RTECS Number ST3000000
Beilstein 13, 120
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2-Nitro-p-phenylenediamine
Packaging Glass bottle
Name Note 95-99%
Assay Percent Range 95-99%
Molecular Formula C6H7N3O2
Linear Formula O2NC6H3(NH2)2
MDL Number MFCD00007903
CAS 5307-14-2
Synonym 1, 4-Diamino-2-nitrobenzene
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Nitro-p-phenylenediamine
Name Note 95-99%
CAS 5307-14-2
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 95-99%
Molecular Formula C6H7N3O2
Linear Formula O2NC6H3(NH2)2
Formula Weight 153.14
MDL Number MFCD00007903
Synonym 1, 4-Diamino-2-nitrobenzene
Accessories

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.

SDS & Safety Documents