missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitro-p-cymene, TCI America™

$227.64 - $776.13

Chemical Identifiers

CAS 943-15-7
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00007176
InChI Key DRKFWQDBPGTSOO-UHFFFAOYSA-N
Synonym 4-Isopropyl-1-methyl-2-nitrobenzene, 4-Isopropyl-2-nitrotoluene
PubChem CID 70340
IUPAC Name 1-methyl-2-nitro-4-propan-2-ylbenzene
SMILES CC1=C(C=C(C=C1)C(C)C)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N08015G View Documents TCI America
N08015G
5g Each for $227.64
 
N080125G missing translation for 'viewDocuments' TCI America
N080125G
25g Each for $776.13
 
Specifications

Chemical Identifiers

943-15-7
179.219
DRKFWQDBPGTSOO-UHFFFAOYSA-N
70340
CC1=C(C=C(C=C1)C(C)C)[N+](=O)[O-]
C10H13NO2
MFCD00007176
4-Isopropyl-1-methyl-2-nitrobenzene, 4-Isopropyl-2-nitrotoluene
1-methyl-2-nitro-4-propan-2-ylbenzene

Specifications

134°C
5g
C10H13NO2
4-Isopropyl-1-methyl-2-nitrobenzene, 4-Isopropyl-2-nitrotoluene
CC1=C(C=C(C=C1)C(C)C)[N+](=O)[O-]
179.219
179.22
≥95.0% (GC)
Yellow
943-15-7
MFCD00007176
DRKFWQDBPGTSOO-UHFFFAOYSA-N
1-methyl-2-nitro-4-propan-2-ylbenzene
70340
Clear Liquid at 20°C
2-Nitro-p-cymene
Safety and Handling

Safety and Handling

TSCA : Yes