missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitro-p-cresol, TCI America™

$33.61 - $302.50

Chemical Identifiers

CAS 119-33-5
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007120
InChI Key SYDNSSSQVSOXTN-UHFFFAOYSA-N
Synonym 2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
PubChem CID 8391
IUPAC Name 4-methyl-2-nitrophenol
SMILES CC1=CC(=C(C=C1)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N018625G View Documents TCI America
N018625G
25g Each for $33.61
 
N0186500G View Documents TCI America
N0186500G
500g Each for $302.50
 
Specifications

Chemical Identifiers

119-33-5
153.137
SYDNSSSQVSOXTN-UHFFFAOYSA-N
8391
CC1=CC(=C(C=C1)O)[N+](=O)[O-]
C7H7NO3
MFCD00007120
2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
4-methyl-2-nitrophenol

Specifications

Undesignated
119-33-5
MFCD00007120
2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
CC1=CC(=C(C=C1)O)[N+](=O)[O-]
153.137
153.14
≥99.0% (GC,T)
25g
C7H7NO3
2446
SYDNSSSQVSOXTN-UHFFFAOYSA-N
4-methyl-2-nitrophenol
8391
Crystal-Lump at 20°C
2-Nitro-p-cresol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0790

RTECSNumber : GP2800000

TSCA : Yes