missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-N-(2-nitrophenyl)benzenesulfonamide, 97%

Chemical Identifiers

CAS 331256-09-8
Molecular Formula C12H9N3O6S
Molecular Weight (g/mol) 323.279
MDL Number MFCD00406329
InChI Key NGHQCODQNBAKLD-UHFFFAOYSA-N
Synonym 2-nitro-n-2-nitrophenyl benzenesulfonamide, n-2-nitrophenylsulfonyl-2-nitroaniline, 2-nitrophenyl 2-nitrophenyl sulfonyl amine, 2-nitro-n-2-nitro-phenyl-benzenesulfonamide
PubChem CID 2827773
IUPAC Name 2-nitro-N-(2-nitrophenyl)benzenesulfonamide
SMILES C1=CC=C(C(=C1)NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60193MD View Documents Alfa Aesar™
H60193MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6019303 View Documents Alfa Aesar™
H6019303
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

331256-09-8
323.279
NGHQCODQNBAKLD-UHFFFAOYSA-N
2827773
C1=CC=C(C(=C1)NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]
C12H9N3O6S
MFCD00406329
2-nitro-n-2-nitrophenyl benzenesulfonamide, n-2-nitrophenylsulfonyl-2-nitroaniline, 2-nitrophenyl 2-nitrophenyl sulfonyl amine, 2-nitro-n-2-nitro-phenyl-benzenesulfonamide
2-nitro-N-(2-nitrophenyl)benzenesulfonamide

Specifications

250mg
C12H9N3O6S
2-nitro-n-2-nitrophenyl benzenesulfonamide, n-2-nitrophenylsulfonyl-2-nitroaniline, 2-nitrophenyl 2-nitrophenyl sulfonyl amine, 2-nitro-n-2-nitro-phenyl-benzenesulfonamide
C1=CC=C(C(=C1)NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]
323.279
323.28
2-Nitro-N-(2-nitrophenyl)benzenesulfonamide
331256-09-8
MFCD00406329
NGHQCODQNBAKLD-UHFFFAOYSA-N
2-nitro-N-(2-nitrophenyl)benzenesulfonamide
2827773
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning