missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitro-m-cresol, TCI America™

$24.44 - $82.50

Chemical Identifiers

CAS 4920-77-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007125
InChI Key QIORDSKCCHRSSD-UHFFFAOYSA-N
Synonym 2-nitro-3-methylphenol, 2-nitro-m-cresol, 2-nitro-3-cresol, m-cresol, 2-nitro, phenol, 3-methyl-2-nitro, 2-hydroxy-6-methyl-nitrobenzene, 2-nitro-3-hydroxytoluene, unii-i4qbb2l3c0, 3-methyl-2-nitro-phenol, 3-hydroxy-2-nitrotoluene
PubChem CID 21026
IUPAC Name 3-methyl-2-nitrophenol
SMILES CC1=C(C(=CC=C1)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N055225G View Documents TCI America
N055225G
25g Each for $82.50
 
N05525G View Documents TCI America
N05525G
5g Each for $24.44
 
Specifications

Chemical Identifiers

4920-77-8
153.137
QIORDSKCCHRSSD-UHFFFAOYSA-N
21026
CC1=C(C(=CC=C1)O)[N+](=O)[O-]
C7H7NO3
MFCD00007125
2-nitro-3-methylphenol, 2-nitro-m-cresol, 2-nitro-3-cresol, m-cresol, 2-nitro, phenol, 3-methyl-2-nitro, 2-hydroxy-6-methyl-nitrobenzene, 2-nitro-3-hydroxytoluene, unii-i4qbb2l3c0, 3-methyl-2-nitro-phenol, 3-hydroxy-2-nitrotoluene
3-methyl-2-nitrophenol

Specifications

108°C
40°C
4920-77-8
MFCD00007125
2-nitro-3-methylphenol, 2-nitro-m-cresol, 2-nitro-3-cresol, m-cresol, 2-nitro, phenol, 3-methyl-2-nitro, 2-hydroxy-6-methyl-nitrobenzene, 2-nitro-3-hydroxytoluene, unii-i4qbb2l3c0, 3-methyl-2-nitro-phenol, 3-hydroxy-2-nitrotoluene
CC1=C(C(=CC=C1)O)[N+](=O)[O-]
153.137
153.14
≥98.0% (GC,T)
Undesignated
25g
C7H7NO3
2446
QIORDSKCCHRSSD-UHFFFAOYSA-N
3-methyl-2-nitrophenol
21026
Crystal-Powder at 20°C
2-Nitro-m-cresol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0790

TSCA : No

Recommended Storage : Refrigerator