Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-9-fluorenone, 99%

Chemical Identifiers

Chemical Name or Material 2-Nitro-9-fluorenone
CAS 3096-52-4
Formula Weight 225.2
MDL Number MFCD00001152
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5569903

 
alfa aesar™
H5569903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5569906

 
alfa aesar™
H5569906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55699MD

 
alfa aesar™
H55699MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.2
Melting Point 222° to 223°C
Chemical Name or Material 2-Nitro-9-fluorenone
Physical Form Solid
Assay 99%
MDL Number MFCD00001152
CAS 3096-52-4
RTECS Number LL9091000
Quantity 1g