Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-5-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Nitro-5-(trifluoromethyl)aniline
CAS 402-14-2
Molecular Formula C7H5F3N2O2
Formula Weight 206.13
MDL Number MFCD00042447
UN Number UN2811
Beilstein 2732344
Synonym 3-Amino-4-nitrobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1231003

  SDS
alfa aesar™
L1231003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1231006

  SDS
alfa aesar™
L1231006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.13
Chemical Name or Material 2-Nitro-5-(trifluoromethyl)aniline
Molecular Formula C7H5F3N2O2
CAS 402-14-2
UN Number UN2811
Beilstein 2732344
Synonym 3-Amino-4-nitrobenzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 97%
MDL Number MFCD00042447
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 98° to 102°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.