Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-5-(1-piperidinyl)phenol, 97%

$29.69 - $370.66

Chemical Identifiers

Chemical Name or Material 2-Nitro-5-(1-piperidinyl)phenol
CAS 157831-75-9
Molecular Formula C11H14N2O3
Formula Weight 224.24
MDL Number MFCD00052663
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2488603

  SDS
alfa aesar™
B2488603
1g Each for $29.69

AAB2488606

  SDS
alfa aesar™
B2488606
5g Each for $95.99

AAB2488614

  SDS
alfa aesar™
B2488614
25g Each for $370.66
Description & Specifications

Specifications

Formula Weight 224.24
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Nitro-5-(1-piperidinyl)phenol
MDL Number MFCD00052663
CAS 157831-75-9
Quantity 1g
Assay 97%
Molecular Formula C11H14N2O3
Melting Point 68° to 70°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.