Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-4-(trifluoromethyl)phenylacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenylacetonitrile
CAS 13544-06-4
Molecular Formula C9H5F3N2O2
Formula Weight 230.15
MDL Number MFCD00045621
UN Number UN3439
Beilstein 2588629
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1923903

 
alfa aesar™
L1923903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1923906

 
alfa aesar™
L1923906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.15
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenylacetonitrile
Molecular Formula C9H5F3N2O2
MDL Number MFCD00045621
CAS 13544-06-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Odor Characteristic
Quantity 1g
Assay 98%
Beilstein 2588629
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Boiling Point 134° to 138°C (0.5mmHg)
Melting Point 38° to 42°C
UN Number UN3439
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.