Please login to your online account to display your discounted pricing

2-Nitro-4-(trifluoromethyl)phenol, 99%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC218880010

 View more versions of this product

Catalog No. AC218880010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 207.11
Quantity 1g
Color Yellow to orange
Density 1.47g/mL
CAS Max % 100.0
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenol
Assay Percent Range 99%
Molecular Formula C7H4F3NO3
Linear Formula O2NC6H3(CF3)OH
CAS 400-99-7
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenol
CAS 400-99-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H4F3NO3
Linear Formula O2NC6H3(CF3)OH
Formula Weight 207.11
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents